Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ Fluoxetine-D6 oxalate Solution, 1.0 mg/mL in Methanol (as free base), Sold by MilliporeSigma™ Supelco™

Catalog No. 01001364 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-001-364 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-001-364 Supplier MilliporeSigma Supelco Supplier No. F0381ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Certified reference material

A Certified Spiking Solution™ suitable as an internal standard for quantitation of Fluoxetine levels in urine or other matrices by LC/MS or GC/MS for pharmaceutical research, clinical toxicology, or forensic testing applications. Fluoxetine is a pharmaceutical antidepressant which functions as a selective serotonin reuptake inhibitor (SSRI). Fluoxetine is sold under the trade name Prozac for treatment of psychological conditions including major depression and obsessive-compulsive disorder.
TRUSTED_SUSTAINABILITY

Specifications

CAS 67-56-1
Linear Formula C17D6H12F3NO · C2H2O4
MDL Number MFCD08276449
UN Number UN 1230 - class 3 - PG 2 - Methanol, solution
Concentration 1.0 mg/mL in methanol (as free base)
Formula Weight 405.40
Packaging Ampule
Quantity 1 mL
Flash Point 9.7°C (49.5°F) - closed cup

CERILLIANT is a registered trademark of Cerilliant Corporation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.