Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ Fluvoxamine maleate, 1.0 mg/mL in Methanol (as free base), Sold by MilliporeSigma™ Supelco™

Catalog No. 01001365 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-001-365 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-001-365 Supplier MilliporeSigma Supelco Supplier No. F0401ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Certified reference material

A Certified Spiking Solution™ of fluvoxamine maleate suitable for use in HPLC/UV or mass spectrometry-based antidepressant testing methods in clinical toxicology, forensic analysis, prescription monitoring or pharmaceutical research applications. Fluvoxamine is a selective serotonin re-uptake inhibitor (SSRI) used primarily for the treatment of obsessive-compulsive disorder (OCD) and major depressive disorder. Fluvoxamine is marketed under brand names such as Faverin, Fevarin, Floxyfral and Luvox.
TRUSTED_SUSTAINABILITY

Specifications

CAS 67-56-1, 61718-82-9
Linear Formula C15H21F3N2O2 · C4H4O4
MDL Number MFCD00269809
UN Number UN 1230 - class 3 - PG 2 - Methanol, solution
Concentration 1.0 mg/mL in methanol (as free base)
Synonym (E)-5-Methoxy-1-[4-(trifluoromethyl)phenyl]-1-pentanone-O-(2-aminoethyl)oxime maleate
Formula Weight 434.41
Packaging Ampule
Quantity 1 mL
Flash Point 9.7°C (49.5°F) - closed cup

CERILLIANT is a registered trademark of Cerilliant Corporation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.