Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ Gabapentin-13C3 Solution, 100 μg/mL in Methanol, Sold by MilliporeSigma™ Supelco™

Catalog No. 01001405 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-001-405 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-001-405 Supplier MilliporeSigma Supelco Supplier No. G0181ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Certified reference material

Sold under the trade names Neurontin™, Nupentin, or Gabarone, gabapentin is an anticonvulsant drug used primarily for treatment of seizures, fibromyalgia, and neuropathic pain. This internal standard is suitable for mass spectrometry-based testing or isotope dilution methods of gabapentin in forensic analysis, clinical toxicology, pain prescription monitoring, or urine drug testing.
TRUSTED_SUSTAINABILITY

Specifications

CAS 67-56-1
Linear Formula C613C3H17NO2
UN Number UN 1230 - class 3 - PG 2 - Methanol
Concentration 100 μg/mL in methanol
Formula Weight 174.21
Packaging Ampule
Quantity 1 mL
Flash Point 9.7°C (49.5°F) - closed cup

CERILLIANT is a registered trademark of Cerilliant Corporation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.