Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ Gabapentin-D10 Solution, 100 μg/mL in Methanol, Sold by MilliporeSigma™ Supelco™

Catalog No. 01001413 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-001-413 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-001-413 Supplier MilliporeSigma Supelco Supplier No. G9011ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Certified reference material

An anticonvulsant, gabapentin is prescribed for treatment of a number of conditions including epilepsy and neuropathic pain. It is sold under the trade names Neurostil, Gabarone, and Nupentin. This stable labeled internal standard is suitable for quantitation of gabapentin levels in women, men or children by LC/MS applications for urine drug testing, pain prescription monitoring, clinical toxicology, or forensic analysis.
TRUSTED_SUSTAINABILITY

Specifications

CAS 67-56-1, 1126623-20-8
Linear Formula C9D10H7NO2
MDL Number MFCD16294730
UN Number UN 1230 - class 3 - PG 2 - Methanol, solution
Concentration 100 μg/mL in methanol
Formula Weight 181.30
Packaging Ampule
Quantity 1 mL
Flash Point 9.7°C (49.5°F) - closed cup

CERILLIANT is a registered trademark of Cerilliant Corporation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.