Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ (+/-)-gamma-Tocopherol-D3 Solution, 500 μg/mL in Methanol, Sold by MilliporeSigma™ Supelco™

Catalog No. 01002167 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-002-167 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-002-167 Supplier MilliporeSigma Supelco Supplier No. V0461ML
 Restricted Item
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Certified reference material

A lipid soluble form of Vitamin E, γ-Tocopherol is important to human health as an antioxidant and food additive. Vitamin E defficiency, often the result of poor nutrition or low absorption of dietary fat, is monitored in patient serum samples by LC-MS/MS or HPLC. This stable-labeled Certified Spiking Solution™ is suitable for quantifying γ-tocopherol in a variety of testing applications from diagnostic testing of Vitamin E defficiency by LC-MS/MS or HPLC to quality control testing of tocopherol-based dietary supplements or pharmaceutical preparations.
TRUSTED_SUSTAINABILITY

Specifications

CAS 67-56-1, 842129-89-9
Linear Formula C28H45D3O2
Concentration 500 μg/mL in methanol
Formula Weight 419.70
Packaging Ampule
Quantity 1 mL
Flash Point 9.7°C (49.5°F) - closed cup

CERILLIANT is a registered trademark of Cerilliant Corporation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.