Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ GHB-d6 sodium salt Solution, 1.0 mg/mL in Methanol (as salt), Sold by MilliporeSigma™ Supelco™

Certified reference material

Supplier:  MilliporeSigma Supelco G0061ML

Catalog No. 01-001-402


 Restricted Item
Add to Cart

Description

Description

GHB, or γ-hydroxybutyrate, is a central nervous system depressant sold under the brand name Xyrem™ and used medically to treat narcolepsy, excessive daytime sleepiness and in rare circumstances, alcoholism. GHB is also used recreationally as an intoxicant and date rape drug. This internal standard is suitable for quantitation of GHB levels in urine, serum, plasma, or oral fluid by LC/MS or GC/MS for urine drug testing, clinical toxicology, forensic analysis, or isotope dilution methods.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

67-56-1 , 362049-53-4
MFCD01861535
1.0 mg/mL in methanol (as salt)
Ampule
9.7°C (49.5°F) - closed cup
C4HNaO3D6
UN 1230 - class 3 - PG 2 - Methanol, solution
132.12
1 mL
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

H331-H370

H311

EINECSNumber : 200-659-6

Recommended Storage : -20°C

SDS
Documents

Documents

Product Certifications
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

CERILLIANT is a registered trademark of Cerilliant Corporation