Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ Hydrocodone Solution, 1.0 mg/mL in Methanol, Sold by MilliporeSigma™ Supelco™

Catalog No. 01001416 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-001-416 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-001-416 Supplier MilliporeSigma Supelco Supplier No. H0031ML
Add to Cart
Add to Cart

Certified reference material

Hydrocodone is a semi-synthetic opioid derived from codeine or thebaine, two naturally-occurring opiates. The drug, also known as dihydrocodeinone, is used as an analgesic to treat pain and as an antitussive for coughing or related conditions. This Certified Spiking Solution™ is suitable as a starting material for calibrators or controls in GC/MS or LC/MS testing applications from pharmaceutical research and pain prescription monitoring to clinical toxicology and forensic analysis.
TRUSTED_SUSTAINABILITY

Specifications

CAS 67-56-1, 125-29-1
Linear Formula C18H21NO3
UN Number UN 1230 - class 3 - PG 2 - Methanol, solution
Concentration 1.0 mg/mL in methanol
Formula Weight 299.36
Packaging Ampule
Quantity 1 mL
Flash Point 9.7°C (49.5°F) - closed cup

CERILLIANT is a registered trademark of Cerilliant Corporation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.