Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ Hydromorphone-3-beta-D-glucuronide, 100 μg/mL in Methanol:Water (1:1), Sold by MilliporeSigma™ Supelco™

Catalog No. 01001430 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-001-430 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-001-430 Supplier MilliporeSigma Supelco Supplier No. H0511ML
Add to Cart
Add to Cart

Certified reference material

Hydromorphone-3-β-D-glucuronide is a primary urinary metabolite of the opioid analgesic hydromorphone. This Certified Snap-N-Spike™ Solution is suitable for use in LC/MS or GC/MS applications for forensic analysis, clinical toxicology, pain prescription monitoring or urine drug testing. Hydromorphone, marketed under brand names including Palladone and Dilaudid™, is a very potent opiate used as an analgesic for treating pain or as a cough suppressant for painful coughing resulting from continued bronchial irritation.
TRUSTED_SUSTAINABILITY

Specifications

CAS 67-56-1
Linear Formula C23H27NO9
MDL Number MFCD00272664
UN Number UN 1993C 3 / PGIII
Concentration 100 μg/mL in methanol:water (1:1)
Synonym Hydromorphone-3-beta-D-glucuronide solution
Formula Weight 461.46
Packaging Ampule
Quantity 1 mL
Flash Point 24°C (75.2°F) - closed cup

CERILLIANT is a registered trademark of Cerilliant Corporation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.