Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ Hydroxyzine diHydrochloride Solution, 1.0 mg/mL in Methanol (as free base), Sold by MilliporeSigma™ Supelco™

Catalog No. 01001464 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-001-464 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-001-464 Supplier MilliporeSigma Supelco Supplier No. H1241ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Certified reference material

Hydroxyzine is a commonly prescribed first generation antihistamine with sedative properties. This drug is a common cocaine adulterant along with lidocaine, phenacetin, and levamisole. This Certified Spiking Solution™ is suitable for use in a variety of GC/MS or LC/MS testing applications in forensic toxicology, urine drug testing, or clinical chemistry.
TRUSTED_SUSTAINABILITY

Specifications

CAS 67-56-1, 2192-20-3
Linear Formula C21H27ClN2O2 · 2HCl
MDL Number MFCD00058200
UN Number UN 1230 - class 3 - PG 2 - Methanol, solution
Concentration 1.0 mg/mL in methanol (as free base)
Formula Weight 447.83
Packaging Ampule
Quantity 1 mL
Flash Point 9.7°C (49.5°F) - closed cup

CERILLIANT is a registered trademark of Cerilliant Corporation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.