Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ Imipramine-D3 maleate Solution, 100 μg/mL in Methanol (as free base), Sold by MilliporeSigma™ Supelco™

Certified reference material

Supplier:  MilliporeSigma Supelco I9031ML

Catalog No. 01-001-512


 Restricted Item
Add to Cart

Description

Description

Imipramine, also known as melipramine, is a tricyclic antidepressant prescribed for the treatment of major depression, enuresis, and anxiety disorder. This internal standard is suitable for quantitation of imipramine levels in urine, serum, or plasma by LC/MS or GC/MS for urine drug testing, clinical toxicology, forensic analysis, or isotope dilution methods. Imipramine is marketed under the trade name Tofranil.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

67-56-1
UN 1230 - class 3 - PG 2 - Methanol, solution
Imipramine-D3 maleate solution
Ampule
9.7°C (49.5°F) - closed cup
C19D3H21N2 · C4H4O4
100 μg/mL in methanol (as free base)
399.50
1 mL
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

H331-H370

H311

EINECSNumber : 200-659-6

Recommended Storage : 2 to 8°C

SDS
Documents

Documents

Product Certifications
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

CERILLIANT is a registered trademark of Cerilliant Corporation