Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ Interference Mix 1 (For Qualitative Use Only), 100-1, Sold by MilliporeSigma™ Supelco™

Catalog No. 01001493 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-001-493 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-001-493 Supplier MilliporeSigma Supelco Supplier No. I0231ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Certified reference material

A multi-component solution standard of routinely monitored over-the-counter (OTC) and prescription drugs: acetaminophen; (-)-cotinine; R,R-(-)-pseudoephedrine; phentermine; caffeine; naproxen; ibuprofen and (-)-nicotine. This standard is for qualitative use only in the evaluation of interferences between the mix components and target analytes of interest in a testing laboratory's analytical methods.
TRUSTED_SUSTAINABILITY

Specifications

CAS 67-56-1
UN Number UN 1230 - class 3 - PG 2 - Methanol, solution
Synonym Qualitative Interference Mixture
Packaging Ampule
Quantity 1 mL
Flash Point 9.7°C (49.5°F) - closed cup

CERILLIANT is a registered trademark of Cerilliant Corporation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.