Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ Interference Mix 5, 100 μg/mL in Methanol (each analyte), Sold by MilliporeSigma™ Supelco™

Certified reference material

Supplier:  MilliporeSigma Supelco I0271ML

Catalog No. 01-001-497


Add to Cart

Description

Description

A multi-component solution standard of routinely monitored over-the-counter (OTC) and prescription drugs carbamazepine, metformin, sertraline, zonisamide, levetiracetam, phenobarbital, topiramate, phenytoin, zolpidem and (R)-phenylephrine. This standard is for qualitative use only in the evaluation of interferences between the mix components and target analytes of interest in a testing laboratory's analytical methods.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

67-56-1
100 μg/mL in methanol (each analyte)
Ampule
9.7°C (49.5°F) - closed cup
UN 1230 - class 3 - PG 2 - Methanol, solution
Qualitative Interference Mixture
1 mL
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

H331-H370

H311

Recommended Storage : -20°C

SDS
Documents

Documents

Product Certifications
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

CERILLIANT is a registered trademark of Cerilliant Corporation