Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ Itraconazole Solution, 2.0 mg/mL (Methanol with 1% 1M HCl), Sold by MilliporeSigma™ Supelco™

Catalog No. 01001488 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-001-488 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-001-488 Supplier MilliporeSigma Supelco Supplier No. I0151ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Certified reference material

Itraconazole is an antifungal drug prescribed for oral or intravenous treatment of fungal infections. The drug is sold under trade names such as Sporanoxor Onmel™. This Certified Spiking Solution™ is suitable as starting material for calibrators, controls, or linearity standards for clinical and diagnostic testing or therapeutic drug monitoring of itraconazole in patient blood, serum, or plasma samples by LC-MS/MS or HPLC.
TRUSTED_SUSTAINABILITY

Specifications

CAS 67-56-1, 7647-01-0
Linear Formula C35H38Cl2N8O4
UN Number UN 3286 8(6.1)(3) / PGII
Concentration 2.0 mg/mL (methanol with 1% 1M HCl)
Formula Weight 705.63
Packaging Ampule
Quantity 1 mL
Flash Point 9.7°C (49.5°F) - closed cup

CERILLIANT is a registered trademark of Cerilliant Corporation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.