Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CERILLIANT™ JWH-073 Solution, 100 μg/mL in Acetonitrile, Sold by MilliporeSigma™ Supelco™
Shop All MilliporeSigma Supelco ProductsDescription
A synthetic cannabinoid found in several versions of the herbal mixture 'Spice', JWH-073 is considerably more potent that natural THC from cannabis and has an extended duration of action. Illicit use centers on its psychoactive effects which mimic those of δ9-THC. This Spice cannabinoid presents significant challenges for detection by typical δ9-THC testing assays. Since the US Drug Enforcement Agency (DEA) listed JWH-073 on its Drugs and Chemicals of Concern list in July, 2009, federal authorities have reported seizures of Spice in Ohio, Pennsylvania, and Florida. Several states have passed legislative action restricting the sale or possession of JWH-073 including Alabama, Arkansas, Kansas, Kentucky, Louisiana, Missouri, and Tennessee.
Specifications
Specifications
| CAS | 75-05-8, 208987-48-8 |
| Linear Formula | C23H21NO |
| MDL Number | MFCD11976912 |
| UN Number | UN 1648 3 / PGII |
| Concentration | 100 μg/mL in acetonitrile |
| Formula Weight | 327.42 |
| Packaging | Ampule |
| Quantity | 1 mL |
| Flash Point | 2°C (35.6°F) - closed cup |
CERILLIANT is a registered trademark of Cerilliant Corporation
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?