Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ Ketamine-D4 Hydrochloride Solution, 1.0 mg/mL in Methanol (as free base), Sold by MilliporeSigma™ Supelco™

Catalog No. 01001524 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-001-524 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-001-524 Supplier MilliporeSigma Supelco Supplier No. K0061ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Certified reference material

This stable-labeled internal standard is suitable for quantitation of ketamine levels in GC/MS or LC/MS applications from clinical toxicology and forensic analysis to urine drug testing and isotope dilution methods. Ketamine, marketed under trade names Ketanest, Ketaset and Ketalar, is an anesthetic used in human and veterinary medicine. The drug is also abused at clubs, raves, or parties for its noted hallucinogenic and dissociative properties and prescribed by some doctors as an off-label treatment for depression.
TRUSTED_SUSTAINABILITY

Specifications

CAS 67-56-1, 1246815-97-3
Linear Formula C13D4H12ClNO · HCl
MDL Number MFCD28038722
UN Number UN 1230 - class 3 - PG 2 - Methanol, solution
Concentration 1.0 mg/mL in methanol (as free base)
Formula Weight 278.21
Packaging Ampule
Quantity 1 mL
Flash Point 9.7°C (49.5°F) - closed cup

CERILLIANT is a registered trademark of Cerilliant Corporation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.