Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ Lamotrigine-13C, 15N4 Solution, 500 μg/mL in Methanol, Sold by MilliporeSigma™ Supelco™

Catalog No. 01001534 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-001-534 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-001-534 Supplier MilliporeSigma Supelco Supplier No. L0221ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Certified reference material

A certified solution and internal standard suitable for GC/MS or LC/MS applications such as clinical toxicology, urine drug testing, forensic analysis or isotope dilution methods. Lamotrigine is an antiepileptic drug marketed under the trade name Lamictal for the treatment of epilepsy and bipolar disorder.
TRUSTED_SUSTAINABILITY

Specifications

CAS 67-56-1
Linear Formula 13CC8H7Cl215N4N
UN Number UN 1230 - class 3 - PG 2 - Methanol, solution
Concentration 500 μg/mL in methanol
Formula Weight 261.06
Packaging Ampule
Quantity 1 mL
Flash Point 9.7°C (49.5°F) - closed cup

CERILLIANT is a registered trademark of Cerilliant Corporation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.