Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ Lamotrigine Solution, 1.0 mg/mL in Methanol, Sold by MilliporeSigma™ Supelco™

Certified reference material

Supplier:  MilliporeSigma Supelco L0191ML

Catalog No. 01-001-531


Add to Cart

Description

Description

Lamotrigine is an anticonvulsant approved for treatment of epilepsy and bipolar disorder. Sold under the trade name Lamictal™, this phenyltriazine analog also is used off-label for treatment of clinical depression. This Snap-N-Spike™ Reference Solution is suitable for use as starting material in calibrators or controls for a variety of LC/MS or GC/MS applications such as forensic analysis and clinical toxicology.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

67-56-1 , 84057-84-1
UN 1230 - class 3 - PG 2 - Methanol, solution
256.09
1 mL
C9H7Cl2N5
1.0 mg/mL in methanol
Ampule
9.7°C (49.5°F) - closed cup
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

H331-H370

H311

Recommended Storage : -20°C

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

CERILLIANT is a registered trademark of Cerilliant Corporation