Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ (-)-Levamisole Hydrochloride Solution, 1.0 mg/mL in Methanol (as free base), Sold by MilliporeSigma™ Supelco™

Certified reference material

Supplier:  MilliporeSigma Supelco L0251ML

Catalog No. 01-001-536


Add to Cart

Description

Description

Levamisole, an antihelminthic drug used to treat worm infestations in humans and animals, is an adulterant and cutting agent found in cocaine sold in the US and Canada. From 2008-2009, levamisole was detected in 69% of cocaine samples seized by the US Drug Enforcement Administration (DEA).
TRUSTED_SUSTAINABILITY
Specifications

Specifications

67-56-1 , 16595-80-5
MFCD00792481
1.0 mg/mL in methanol (as free base)
Ampule
9.7°C (49.5°F) - closed cup
C11H12N2S
UN 1230 - class 3 - PG 2 - Methanol, solution
204.29
1 mL
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

H331-H370

H311

Recommended Storage : -20°C

SDS
Documents

Documents

Product Certifications
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

CERILLIANT is a registered trademark of Cerilliant Corporation