Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ Lidocaine-D10 Solution, 100 μg/mL in Methanol, Sold by MilliporeSigma™ Supelco™

Catalog No. 01001554 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-001-554 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-001-554 Supplier MilliporeSigma Supelco Supplier No. L0501ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Certified reference material

Lidocaine is a local anesthetic and antiarrhythmic drug. Lidocaine is often added to cocaine as a diluent. A Certified Snap-N-Spike™ Solution for use in clinical toxicology, forensic analysis, urine drug testing, or pharmaceutical research applications by LC/MS or GC/MS.
TRUSTED_SUSTAINABILITY

Specifications

CAS 67-56-1, 851528-09-1
Linear Formula C14H12D10N2O
Concentration 100 μg/mL in methanol
Formula Weight 244.40
Packaging Ampule
Quantity 1 mL
Flash Point 9.7°C (49.5°F) - closed cup

CERILLIANT is a registered trademark of Cerilliant Corporation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.