Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ Lisdexamfetamine-D4 dimesylate Solution, 100 μg/mL in Methanol (as free base), Sold by MilliporeSigma™ Supelco™

Catalog No. 01001539 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-001-539 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-001-539 Supplier MilliporeSigma Supelco Supplier No. L0281ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Certified reference material

A certified solution standard suitable for quantitation of lisdexamfetamine in urine or other biological samples by GC/MS or LC/MS in clinical toxicology testing, forensic analysis, urine drug testing, or pharmaceutical research. Lisdexamfetamine, a psychostimulant prodrug of dextroamphetamine, is marketed as the drug Vyvanse™ for treatment of attention deficit hyperactivity disorder (ADHD) in adults and children six to twelve years of age.
TRUSTED_SUSTAINABILITY

Specifications

CAS 67-56-1
Linear Formula C15H21D4N3O · C2H6S2O6H2
UN Number UN 1230 - class 3 - PG 2 - Methanol, solution
Concentration 100 μg/mL in methanol (as free base)
Synonym Lisdexamfetamine-D4 dimesylate solution
Formula Weight 459.61
Packaging Ampule
Quantity 1 mL
Flash Point 9.7°C (49.5°F) - closed cup

CERILLIANT is a registered trademark of Cerilliant Corporation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.