Learn More
CERILLIANT™ Loprazolam Solution, 1 mg/mL (Acetonitrile:DMSO (90:10) (v/v)), Sold by MilliporeSigma™ Supelco™
Certified reference material
Supplier:  MilliporeSigma Supelco L0491ML
Catalog No. 01-001-553
Add to Cart
Description
Description
Loprazolam, marketed under the brand names Dormonoct, Havlane, Sonin and Somnovit, is an imidazolobenzodiazepine derivative with anxiolytic, anticonvulsant, hypnotic, sedative and skeletal muscle relaxant properties. It is licensed and marketed for the short-term treatment of moderately-severe insomnia. This Certified Spiking Solution™ is suitable for use as starting material in calibrators or controls for a variety of LC/MS or GC/MS applications from forensic analysis and clinical toxicology to urine drug testing.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
75-05-8 , 61197-73-7
1 mg/mL (acetonitrile:DMSO (90:10) (v/v))
Ampule
2°C (35.6°F) - closed cup
C23H21ClN6O3
464.90
1 mL
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
H332-H319
H312
Recommended Storage : -20°C
SDS
Documents
Documents
Product Certifications
Product Title
Select an issue
CERILLIANT is a registered trademark of Cerilliant Corporation