Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ Lorazepam glucuronide Solution, 100 μg/mL in Acetonitrile:Water (1:1), Sold by MilliporeSigma™ Supelco™

Catalog No. 01001533 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-001-533 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-001-533 Supplier MilliporeSigma Supelco Supplier No. L0211ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Certified reference material

Lorazepam glucuronide is a primary urinary metabolite of lorazepam, a benzodiazepine often prescribed for treatment of anxiety disorders. This Certified Spiking Solution™ is suitable for use as starting material in calibrators or controls for a variety of LC/MS or GC/MS applications from clinical toxicology and forensic analysis to urine drug testing.
TRUSTED_SUSTAINABILITY

Specifications

CAS 75-05-8, 32781-79-6
Linear Formula C21H18Cl2N2O8
MDL Number MFCD02682126
UN Number UN 1648 3 / PGII
Concentration 100 μg/mL in acetonitrile:water (1:1)
Formula Weight 497.28
Packaging Ampule
Quantity 1 mL
Flash Point 13°C (55.4°F) - closed cup

CERILLIANT is a registered trademark of Cerilliant Corporation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.