Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ Loxapine succinate Solution, 1 mg/mL in Methanol (as free base), Sold by MilliporeSigma™ Supelco™

Catalog No. 01001556 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-001-556 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-001-556 Supplier MilliporeSigma Supelco Supplier No. L0521ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Certified reference material

A Certified Snap-N-Spike™ Solution for use in clinical toxicology, forensic analysis, or urine drug testing applications by LC/MS or GC/MS. Loxapine, structurally related to clozapine, is a typical antipsychotic drug used primarily in the treatment of schizophrenia. Loxapine may be metabolized to amoxapine, a tricyclic antidepressant.
TRUSTED_SUSTAINABILITY

Specifications

CAS 67-56-1, 27833-64-3
Linear Formula C18H18ClN3O · C4H6O4
MDL Number MFCD00069298
Concentration 1 mg/mL in methanol (as free base)
Formula Weight 445.90
Packaging Ampule
Quantity 1 mL
Flash Point 9.7°C (49.5°F) - closed cup

CERILLIANT is a registered trademark of Cerilliant Corporation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.