Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ LSD Solution, 25 μg/mL in Acetonitrile, Sold by MilliporeSigma™ Supelco™

Catalog No. 01001528 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-001-528 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-001-528 Supplier MilliporeSigma Supelco Supplier No. L0051ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Certified reference material

LSD, or lysergic acid diethylamide, is an illicit psychedelic drug with potent hallucinogenic effects such as altered thinking processes, closed- and open-eye visuals, synesthesia and an altered sense of time. This Snap-N-Spike™ reference solution is suitable for use in urine drug testing, clinical toxicology, or forensic analysis by LC-MS/MS or GC/MS.
TRUSTED_SUSTAINABILITY

Specifications

CAS 75-05-8, 50-37-3
Linear Formula C20H25N3O
MDL Number MFCD00135795
UN Number UN 1648 3 / PGII
Concentration 25 μg/mL in acetonitrile
Synonym Lysergic acid diethylamide
Formula Weight 323.43
Packaging Ampule
Quantity 1 mL
Flash Point 2°C (35.6°F) - closed cup

CERILLIANT is a registered trademark of Cerilliant Corporation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.