Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ MAB-CHMINACA Solution, 100 μg/mL in Methanol, Sold by MilliporeSigma™ Supelco™

Catalog No. 01002005 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-002-005 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-002-005 Supplier MilliporeSigma Supelco Supplier No. S1091ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Certified reference material

MAB-CHMINACA, also known as ADB-CHMINACA, is a synthetic cannabinoid found on the illicit drug market. This certified Snap-N-Spike solution is suitable as starting material for calibrators or controls in synthetic cannabinoid GC/MS or LC/MS methods for urine drug testing, forensic analysis, or clinical toxicology applications. For those synthetic cannabinoids currently controlled at the federal level, our certified solution standards of these substances are DEA-exempt, allowing laboratories to place orders without additional regulatory paperwork. For US customers, state specific regulatory requirements may apply.
TRUSTED_SUSTAINABILITY

Specifications

CAS 67-56-1, 1863065-92-2
Linear Formula C21H30N4O2
MDL Number MFCD30718192
UN Number UN 1230 - class 3 - PG 2 - Methanol, solution
Concentration 100 μg/mL in methanol
Formula Weight 370.49
Packaging Ampule
Quantity 1 mL
Flash Point 9.7°C (49.5°F) - closed cup

CERILLIANT is a registered trademark of Cerilliant Corporation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.