Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ Maprotiline Hydrochloride Solution, 1.0 mg/mL in Methanol (as free base), Sold by MilliporeSigma™ Supelco™

Certified reference material

Supplier:  MilliporeSigma Supelco M9201ML

Catalog No. 01-001-698


Add to Cart

Description

Description

Maprotiline is a tetracyclic antidepressant marketed under the trade name Ludiomil for treatment of depression. This Certified Spiking Solution™ suitable for use in urine drug testing, clinical toxicology, or forensic analysis by LC-MS/MS or GC/MS.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

67-56-1 , 10347-81-6
UN 1230 - class 3 - PG 2 - Methanol, solution
313.86
1 mL
C20H24ClN
1.0 mg/mL in methanol (as free base)
Ampule
9.7°C (49.5°F) - closed cup
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

H331-H370

H311

EINECSNumber : 200-659-6

Recommended Storage : 2 to 8°C

SDS
Documents

Documents

Product Certifications
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

CERILLIANT is a registered trademark of Cerilliant Corporation