Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ (+/-)-MDEA-D6 Solution, 1.0 mg/mL in Methanol, Sold by MilliporeSigma™ Supelco™

Catalog No. 01001605 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-001-605 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-001-605 Supplier MilliporeSigma Supelco Supplier No. M0821ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Certified reference material

MDEA is an illicit recreational drug of the amphetamine and phenethylamine classes. This internal standard is suitable for MDEA testing or isotope dilution methods by GC/MS or LC/MS for clinical toxicology, urine drug testing, or forensic analysis. A 3,4-methylenedioxy derivative of N-ethylamphetamine, MDEA is abused for its psychedelic effects, similar to those of its structural analog MDMA (Ecstasy).
TRUSTED_SUSTAINABILITY

Specifications

CAS 67-56-1, 160227-44-1
Linear Formula C12D6H11NO2
UN Number UN 1230 - class 3 - PG 2 - Methanol, solution
Concentration 1.0 mg/mL in methanol
Synonym (+/-)-3,4-Methylenedioxyethylamphetamine-d6
Formula Weight 213.31
Packaging Ampule
Quantity 1 mL
Flash Point 9.7°C (49.5°F) - closed cup

CERILLIANT is a registered trademark of Cerilliant Corporation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.