Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ (+/-)-MDMA-D5 Solution, 100 μg/mL in Methanol, Sold by MilliporeSigma™ Supelco™

Catalog No. 01001570 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-001-570 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-001-570 Supplier MilliporeSigma Supelco Supplier No. M0111ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Certified reference material

MDMA is an illicit drug known by a variety of street names, including 'Ecstasy,' 'E,' 'X,' and 'XTC'. An analog of the phenethylamine and amphetamine classes, MDMA is abused for its euphoric and psychedelic effects. This stable-labeled internal standard is suitable for quantitation of MDMA levels in urine, serum, or plasma by LC/MS or GC/MS for urine drug testing, clinical toxicology, forensic analysis, or isotope dilution methods.
TRUSTED_SUSTAINABILITY

Specifications

CAS 67-56-1, 136765-43-0
Linear Formula C11D5H10NO2
UN Number UN 1230 - class 3 - PG 2 - Methanol, solution
Concentration 100 μg/mL in methanol
Synonym (+/-)-3,4-Methylenedioxymethamphetamine-d5
Formula Weight 198.27
Packaging Ampule
Quantity 1 mL
Flash Point 9.7°C (49.5°F) - closed cup

CERILLIANT is a registered trademark of Cerilliant Corporation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.