Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ (+/-)-MDMA Solution, 1.0 mg/mL in Methanol, Sold by MilliporeSigma™ Supelco™

Catalog No. 01001572 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-001-572 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-001-572 Supplier MilliporeSigma Supelco Supplier No. M0131ML
 Restricted Item
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Certified reference material

MDMA is an illicit drug known by a variety of street names, including 'Ecstasy,' 'E,' 'X,' and 'XTC'. An analog of the phenethylamine and amphetamine classes, MDMA is abused for its euphoric and psychedelic effects. This certified Snap-N-Spike™ Solution is suitable for use as starting material in calibrators or controls for a variety of LC/MS or GC/MS applications from forensic analysis and clinical toxicology to urine drug testing.
TRUSTED_SUSTAINABILITY

Specifications

CAS 67-56-1, 42542-10-9
Linear Formula C11H15NO2
MDL Number MFCD00287291
UN Number UN 1230 - class 3 - PG 2 - Methanol, solution
Concentration 1.0 mg/mL in methanol
Synonym (+/-)-3,4-Methylenedioxymethamphetamine
Formula Weight 193.24
Packaging Ampule
Quantity 1 mL
Flash Point 9.7°C (49.5°F) - closed cup

CERILLIANT is a registered trademark of Cerilliant Corporation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.