Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ MDMB-FUBINACA Solution, 100 μg/mL in Acetonitrile, Sold by MilliporeSigma™ Supelco™

Catalog No. 01002011 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-002-011 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-002-011 Supplier MilliporeSigma Supelco Supplier No. S1171ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Certified reference material

MDMB-FUBINACA, also known as MDMB(N)-Bz-F and FUB-MDMB, is a potent indazole-based synthetic cannabinoid receptor agonist sold as a designer drug. This Certified Spiking Solution™ is suited for use in synthetic cannabinoid testing methods by LC/MS or GC/MS for applications in clinical toxicology, urine drug testing, and forensic analysis. For those synthetic cannabinoids currently controlled at the federal level, our certified solution standards of these substances are DEA-exempt, allowing laboratories to place orders without additional regulatory paperwork. For US customers, state specific regulatory requirements may apply.
TRUSTED_SUSTAINABILITY

Specifications

CAS 75-05-8, 1971007-93-8
Linear Formula C22H24FN3O3
Concentration 100 μg/mL in acetonitrile
Formula Weight 397.44
Packaging Ampule
Quantity 1 mL
Flash Point 2°C (35.6°F) - closed cup

CERILLIANT is a registered trademark of Cerilliant Corporation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.