Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ MDPBP Hydrochloride Solution, 1.0 mg/mL in Methanol (as free base), Sold by MilliporeSigma™ Supelco™

Catalog No. 01001641 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-001-641 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-001-641 Supplier MilliporeSigma Supelco Supplier No. M1541ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Certified reference material

A novel designer drug first developed in the 1960's as a research chemical, MDPBP first emerged as a legal high in Europe in 2009. This synthetic cathinone and bath salt is often sold in the US and other countries under the name 'NRG-1' as a mixture with other substituted cathinone derivatives such as flephedrone, pentylone, and MDPV.
TRUSTED_SUSTAINABILITY

Specifications

CAS 67-56-1, 24622-60-4
Linear Formula C15H19NO3 · HCl
MDL Number MFCD27966833
UN Number UN 1230 - class 3 - PG 2 - Methanol, solution
Concentration 1.0 mg/mL in methanol (as free base)
Synonym 3′,4′-Methylenedioxy-alpha-pyrrolidinobutiophenone hydrochloride solution, 3′,4′-Methylenedioxy-alpha-pyrrolidinobutiophenone hydrochloride solution
Formula Weight 297.78
Packaging Ampule
Quantity 1 mL
Flash Point 9.7°C (49.5°F) - closed cup

CERILLIANT is a registered trademark of Cerilliant Corporation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.