Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ Medazepam Solution, 1 mg/mL in Methanol, Sold by MilliporeSigma™ Supelco™

Catalog No. 01001678 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-001-678 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-001-678 Supplier MilliporeSigma Supelco Supplier No. M2051ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Certified reference material

Medazepam, marketed as Azepamid, Nobrium, Tranquirax, Rudotel, Raporan, Ansilan and Mezapam, is a long-acting benzodiazepine drug used as anxiolytic, anticonvulsant, sedative, and skeletal muscle relaxant. This certified solution standard is suitable for numerous GC/MS or LC/MS testing applications in forensic analysis, clinical toxicology, urine drug testing, or therapeutic drug monitoring.
TRUSTED_SUSTAINABILITY

Specifications

CAS 67-56-1, 2898-12-6
Linear Formula C16H15ClN2
MDL Number MFCD00063407
UN Number UN 1230 - class 3 - PG 2 - Methanol, solution
Concentration 1 mg/mL in methanol
Formula Weight 270.76
Packaging Ampule
Quantity 1 mL
Flash Point 11°C (51.8°F) - closed cup

CERILLIANT is a registered trademark of Cerilliant Corporation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.