Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CERILLIANT™ Meperidine-D4 Solution, 100 μg/mL in methanol, Sold by MilliporeSigma™ Supelco™
Shop All MilliporeSigma Supelco ProductsSpecifications
Specifications
| Packaging | Ampoule |
| CAS | 53484-73-4 |
| Product Line | CERILLIANT |
| Physical Form | Liquid |
| Molecular Formula | C15D4H17NO2 |
| Concentration | 100 μg/mL in Methanol |
| Industry Type | Forensics, Toxicology |
| Molecular Weight (g/mol) | 251.36 |
| Quantity | 1 mL |
| Grade | Certified Reference Material |
CERILLIANT™ is a registered trademark of Cerilliant Corporation
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?