Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ Meprobamate-13C3 Solution, 100 μg/mL in Methanol, Sold by MilliporeSigma™ Supelco™

Certified reference material

Supplier:  MilliporeSigma Supelco M1781ML

Catalog No. 01-001-657


Add to Cart

Description

Description

This stable-labeled internal standard is suitable for meprobamate testing or isotope dilution methods by LC/MS or GC/MS for clinical toxicology, forensic analysis, pain prescription monitoring, or urine drug testing applications. Marketed under trade names such as Milltown, Meprospan, and Equanil, meprobamate is a minor tranquilizer used for the treatment of anxiety and muscle spasms.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

75-05-8
C613C3H18N2O4
100 μg/mL in methanol
221.23
1 mL
-47.99°C
UN 1648 3 / PGII
0.78 g/cm3 at 20°C
Ampule
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

H332-H319

H312

EINECSNumber : 200-835-2

Recommended Storage : -20°C

SDS
Documents

Documents

Product Certifications
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

CERILLIANT is a registered trademark of Cerilliant Corporation