Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ (+/-)-Metanephrine-D3 Hydrochloride Solution, 100 μg/mL in Methanol, Sold by MilliporeSigma™ Supelco™

Catalog No. 01001637 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-001-637 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-001-637 Supplier MilliporeSigma Supelco Supplier No. M1481ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Certified reference material

Measurement of metanephrine, a metabolite of epinephrine, is a useful tool in the diagnosis of diseases such as pheochromocytoma and adrenal medullary neoplasm. This internal standard is suitable for LC-MS/MS monitoring of metanephrine levels in plasma or urine for diagnostic testing, endocrinology, or clinical chemistry.
TRUSTED_SUSTAINABILITY

Specifications

CAS 67-56-1, 1215507-88-2
Linear Formula C10H12D3NO3·HCl
UN Number UN 1230 - class 3 - PG 2 - Methanol, solution
Concentration 100 μg/mL in methanol (as free base)
Synonym (+/-)-Metanephrine-D3 HCl
Formula Weight 236.71
Packaging Ampule
Quantity 1 mL
Flash Point 9.7°C (49.5°F) - closed cup

CERILLIANT is a registered trademark of Cerilliant Corporation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.