Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ Metaxalone Solution, 1.0 mg/mL in Methanol, Sold by MilliporeSigma™ Supelco™

Certified reference material

Supplier:  MilliporeSigma Supelco M0741ML

Catalog No. 01-001-599


Add to Cart

Description

Description

Metaxalone is a muscle relaxant sold under the trade name Skelaxin™ for relief of muscle pain. This Certified Snap-N-Spike™ Solution is applicable for use in LC/MS or GC/MS applications for pain prescription monitoring, urine drug testing, clinical toxicology and forensic analysis.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

67-56-1 , 1665-48-1
UN 1230 - class 3 - PG 2 - Methanol, solution
221.25
1 mL
C12H15NO3
1.0 mg/mL in methanol
Ampule
9.7°C (49.5°F) - closed cup
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

H331-H370

H311

EINECSNumber : 200-659-6

Recommended Storage : -20°C

SDS
Documents

Documents

Product Certifications
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

CERILLIANT is a registered trademark of Cerilliant Corporation