Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ Methamphetamine/Cocaine/Heroin Solution, 250 μg/mL each component in acetonitrile, Sold by MilliporeSigma™ Supelco™

Catalog No. 01258401 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-258-401 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-258-401 Supplier MilliporeSigma Supelco Supplier No. M0251ML
 Restricted Item
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

A certified Snap-N-Spike™ reference solution of routinely tested drugs methamphetamine, cocaine, and heroin. This analytical reference standard is applicable in LC/MS or GC/MS applications for urine drug testing, clinical toxicology, and forensic analysis.

TRUSTED_SUSTAINABILITY

Specifications

Packaging Ampoule
Product Line CERILLIANT
Physical Form Liquid
Concentration 250 μg/mL each component in Acetonitrile
Industry Type Forensics, Toxicology
Quantity 1 mL
Grade Certified Reference Material

CERILLIANT™ is a registered trademark of Cerilliant Corporation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.