Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ (+/-)-Methamphetamine-D11 Solution, 1.0 mg/mL in Methanol, Sold by MilliporeSigma™ Supelco™

Catalog No. 01001593 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-001-593 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-001-593 Supplier MilliporeSigma Supelco Supplier No. M0601ML
Add to Cart
Add to Cart

Certified reference material

Methamphetamine is a psychostimulant drug of the phenethylamine and amphetamine classes known by the street names 'meth,' 'ice,' and 'glass'. This internal standard is suitable for quantitation of methamphetamine levels in urine, serum, or plasma by LC/MS or GC/MS for urine drug testing, clinical toxicology, sports testing, forensic analysis, or isotope dilution methods. The dextrorotatory isomer of methamphetamine is a pharmaceutical drug sold under the trade name Desoxyn™ for treatment of ADHD and obesity.
TRUSTED_SUSTAINABILITY

Specifications

CAS 67-56-1, 152477-88-8
Linear Formula C10D11H4N
MDL Number MFCD00673293
UN Number UN 1230 - class 3 - PG 2 - Methanol, solution
Concentration 1.0 mg/mL in methanol
Formula Weight 160.30
Packaging Ampule
Quantity 1 mL
Flash Point 9.7°C (49.5°F) - closed cup

CERILLIANT is a registered trademark of Cerilliant Corporation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.