Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ (+/-)-Methamphetamine-D11 Solution, 100 μg/mL in Methanol, Sold by MilliporeSigma™ Supelco™

Certified reference material

Supplier:  MilliporeSigma Supelco M0591ML

Catalog No. 01-001-592


Add to Cart

Description

Description

Methamphetamine is a psychostimulant drug of the phenethylamine and amphetamine classes known by the street names 'meth,' 'ice,' and 'glass'. This internal standard is suitable for quantitation of methamphetamine levels in urine, serum, or plasma by LC/MS or GC/MS for urine drug testing, clinical toxicology, sports testing, forensic analysis, or isotope dilution methods. The dextrorotatory isomer of methamphetamine is a pharmaceutical drug sold under the trade name Desoxyn™ for treatment of ADHD and obesity.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

67-56-1 , 152477-88-8
MFCD00673293
100 μg/mL in methanol
Ampule
9.7°C (49.5°F) - closed cup
C10D11H4N
UN 1230 - class 3 - PG 2 - Methanol, solution
160.30
1 mL
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

H331-H370

H311

EINECSNumber : 200-659-6

Recommended Storage : 2 to 8°C

SDS
Documents

Documents

Product Certifications
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

CERILLIANT is a registered trademark of Cerilliant Corporation