Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ (+/-)-Methcathinone-D3 Hydrochloride Solution, 100 μg/mL in Acetonitrile, Sold by MilliporeSigma™ Supelco™

Catalog No. 01001665 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-001-665 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-001-665 Supplier MilliporeSigma Supelco Supplier No. M1891ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Certified reference material

An internal standard suitable for quantification of methcathinone in novel psychoactive substance (NPS) testing methods by LC/MS or GC/MS for applications in forensic analysis, urine drug testing, or clinical toxicology. Known by street names 'Em-kat' and 'Khat', methcathinone is an illicit drug with stimulant and euphoric effects. This drug has also been found in products marketed as bath salts.
TRUSTED_SUSTAINABILITY

Specifications

CAS 75-05-8, 1246819-12-4
Linear Formula C10H10D3NO · HCl
UN Number UN 1648 3 / PGII
Concentration 100 μg/mL in acetonitrile (as free base)
Formula Weight 202.70
Packaging Ampule
Quantity 1 mL
Flash Point 2°C (35.6°F) - closed cup

CERILLIANT is a registered trademark of Cerilliant Corporation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.