Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ Methylmalonic acid Solution, 1.0 mg/mL in Acetonitrile, Sold by MilliporeSigma™ Supelco™

Catalog No. 01001603 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-001-603 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-001-603 Supplier MilliporeSigma Supelco Supplier No. M0801ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Certified reference material

Methylmalonic acid is an indicator of vitamin B12 defficiency and methylmalonic acidemia. This Certified Spiking Solution™ is suitable for use in preparation of calibrators or controls in clinical and diagnostic testing using GC/MS or LC/MS.
TRUSTED_SUSTAINABILITY

Specifications

CAS 75-05-8, 516-05-2
Linear Formula C4H6O4
UN Number UN 1648 3 / PGII
Concentration 1.0 mg/mL in acetonitrile
Formula Weight 118.09
Packaging Ampule
Quantity 1 mL
Flash Point 2°C (35.6°F) - closed cup

CERILLIANT is a registered trademark of Cerilliant Corporation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.