Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ Methylone-D3 Hydrochloride Solution, 100 μg/mL in Methanol (as free base), Sold by MilliporeSigma™ Supelco™

Certified reference material

Supplier:  MilliporeSigma Supelco M1411ML

Catalog No. 01-001-631


Add to Cart

Description

Description

A new internal standard suitable for numerous GCMS and LCMS applications from clinical toxicology testing and forensic analysis to urine drug testing and isotope dilution methods. Also known as bk-MDMA or 3,4-methylenedioxy-N-methylcathinone, methylone is a synthetic stimulant of the amphetamine and cathinone classes with significant abuse reported in the UK, Netherlands, Sweden, and New Zealand.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

67-56-1 , 1246820-21-2
UN 1230 - class 3 - PG 2 - Methanol, solution
246.71
1 mL
C11D3H10NO3 · HCl
100 μg/mL in methanol (as free base)
Ampule
9.7°C (49.5°F) - closed cup
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

H331-H370

H311

EINECSNumber : 200-659-6

Recommended Storage : -20°C

SDS
Documents

Documents

Product Certifications
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

CERILLIANT is a registered trademark of Cerilliant Corporation