Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ Methylphenidate Hydrochloride Solution, (Racemic mixture), Sold by MilliporeSigma™ Supelco™

Catalog No. 01001606 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-001-606 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-001-606 Supplier MilliporeSigma Supelco Supplier No. M0831ML
 Restricted Item
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Certified reference material

Methylphenidate is a psychostimulant prescribed for treatment of attention-deficit hyperactivity disorder (ADHD), tachycardia and narcolepsy. This Certified Spiking Solution™ is applicable for use in LC/MS or GC/MS applications for urine drug testing, clinical toxicology and forensic analysis. Methylphenidate is sold under trade names Concerta™ and Ritalin™.
TRUSTED_SUSTAINABILITY

Specifications

CAS 67-56-1, 298-59-9
Linear Formula C14H20ClNO2
MDL Number MFCD00058191
UN Number UN 1230 - class 3 - PG 2 - Methanol, solution
Formula Weight 269.77
Packaging Ampule
Quantity 1 mL
Flash Point 9.7°C (49.5°F) - closed cup

CERILLIANT is a registered trademark of Cerilliant Corporation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.