Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ Methylphenobarbital, 1.0 mg/mL in Methanol, Sold by MilliporeSigma™ Supelco™

Catalog No. 01001667 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-001-667 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-001-667 Supplier MilliporeSigma Supelco Supplier No. M1931ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Certified reference material

Mephobarbital, marketed under brand names such as Mebaral, Mephyltaletten, and Phemiton, is a long-acting barbiturate medically used as an anticonvulsant and also as a sedative. This Certified Spiking Solution™ is suitable for numerous GC/MS or LC/MS testing applications from clinical toxicology to urine drug testing and forensic analysis.
TRUSTED_SUSTAINABILITY

Specifications

CAS 67-56-1, 115-38-8
Linear Formula C13H14N2O3
MDL Number MFCD00057563
UN Number UN 1230 - class 3 - PG 2 - Methanol, solution
Concentration 1.0 mg/mL in methanol
Formula Weight 246.26
Packaging Ampule
Quantity 1 mL
Flash Point 9.7°C (49.5°F) - closed cup

CERILLIANT is a registered trademark of Cerilliant Corporation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.