Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ Metoprolol tartrate Solution, 1.0 mg/mL in Methanol (as free base), Sold by MilliporeSigma™ Supelco™

Catalog No. 01001620 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-001-620 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-001-620 Supplier MilliporeSigma Supelco Supplier No. M1231ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Certified reference material

A certified solution standard applicable for use in clinical toxicology or forensic analysis by LC-MS/MS or GC/MS. Metoprolol is used for the treatment of multiple heart conditions including hypertension, angina, and tachycardia. This beta blocker is sold under the trade names Lepressor and Toprol XL™.
TRUSTED_SUSTAINABILITY

Specifications

CAS 67-56-1, 56392-17-7
Linear Formula C15H25NO3 · 0.5 C4H6O6
MDL Number MFCD23141017
UN Number UN 1230 - class 3 - PG 2 - Methanol, solution
Concentration 1.0 mg/mL in methanol (as free base)
Formula Weight 342.41
Packaging Ampule
Quantity 1 mL
Flash Point 9.7°C (49.5°F) - closed cup

CERILLIANT is a registered trademark of Cerilliant Corporation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.