Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ Milnacipran Hydrochloride Solution, 1.0 mg/mL in Methanol (as free base), Sold by MilliporeSigma™ Supelco™

Catalog No. 01001634 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-001-634 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-001-634 Supplier MilliporeSigma Supelco Supplier No. M1451ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Certified reference material

Milnacipran, sold under the brand names Savella™, Ixel™, Dalcipran™, or Toledomin, is a serotonin-norepinephrine reuptake inhibitor (SNRI) used in the treatment of depression. In 2009, the US Food and Drug Administration (FDA) approved milnacipran for management of the chronic pain disorder fibromyalgia, making it only the third drug approved in the US for this indication besides duloxetine and pregabalin.
TRUSTED_SUSTAINABILITY

Specifications

CAS 67-56-1, 101152-94-7
Linear Formula C15H23ClN2O
MDL Number MFCD27978092
UN Number UN 1230 - class 3 - PG 2 - Methanol, solution
Concentration 1.0 mg/mL in methanol (as free base)
Formula Weight 282.81
Packaging Ampule
Quantity 1 mL
Flash Point 9.7°C (49.5°F) - closed cup

CERILLIANT is a registered trademark of Cerilliant Corporation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.