Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ Morphine-3-beta-D-glucuronide Solution, 100 μg/mL in Methanol:Water ( 50:50(v/v)), Sold by MilliporeSigma™ Supelco™

Catalog No. 01001575 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-001-575 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-001-575 Supplier MilliporeSigma Supelco Supplier No. M0181ML
 Restricted Item
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Certified reference material

A certified solution standard of Morphine-3-β-D-glucuronide at a concentration of 100 μg/mL for low volumes of sample testing. This certified solution standard is suitable for LC/MS or GC/MS applications including forensic analysis or urine drug testing.
TRUSTED_SUSTAINABILITY

Specifications

CAS 67-56-1, 20290-09-9
Linear Formula C23H27NO9
MDL Number MFCD00056773
UN Number UN 1993C 3 / PGIII
Concentration 100 μg/mL in methanol:water ( 50:50(v/v))
Formula Weight 461.46
Packaging Ampule
Quantity 1 mL
Flash Point 24°C (75.2°F) - closed cup

CERILLIANT is a registered trademark of Cerilliant Corporation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.