Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ Morphine-6-beta-D-glucuronide Solution, 1.0 mg/mL in Methanol:Water (2:8), Sold by MilliporeSigma™ Supelco™

Certified reference material

Supplier:  MilliporeSigma Supelco M0461ML

Catalog No. 01-001-588


Add to Cart

Description

Description

A certified solution standard of Morphine-6-β-D-glucuronide at a concentration of 1.0 mg/mL for high volumes of sample testing. This new certified solution standard is suitable for LCMS or GCMS applications including forensic analysis or urine drug testing.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

67-56-1
MFCD00079434
1.0 mg/mL in methanol:water (2:8)
Ampule
43°C (109.4°F) - closed cup
C23H27NO9
UN 1993C 3 / PGIII
461.46
1 mL
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

H332-H370

H312

Recommended Storage : -20°C

SDS
Documents

Documents

Product Certifications
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

CERILLIANT is a registered trademark of Cerilliant Corporation