Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ Morphine-D3 Solution, 1.0 mg/mL in Methanol, Sold by MilliporeSigma™ Supelco™

Catalog No. 01001566 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-001-566 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-001-566 Supplier MilliporeSigma Supelco Supplier No. M0061ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Certified reference material

This stable-labeled internal standard is suitable for quantiation of morphine levels in urine, serum, or plasma by LC/MS or GC/MS for urine drug testing, pain prescription monitoring, clinical toxicology, forensic analysis, or isotope dilution methods. Morphine is a very potent opiate analgesic drug used to relieve both acute and chronic severe pain. Sold under trade names including MS Contin™, Kadian™, Oramorph™, and Roxanol, morphine is also a commonly abused recreational drug due to its widespread availability and high potential for addiction.
TRUSTED_SUSTAINABILITY

Specifications

CAS 67-56-1, 67293-88-3
Linear Formula C17H16NO3D3
UN Number UN 1230 - class 3 - PG 2 - Methanol, solution
Concentration 1.0 mg/mL in methanol
Formula Weight 288.36
Packaging Ampule
Quantity 1 mL
Flash Point 9.7°C (49.5°F) - closed cup

CERILLIANT is a registered trademark of Cerilliant Corporation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.