Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ Morphine Solution, 100 μg/mL in Methanol, Sold by MilliporeSigma™ Supelco™

Catalog No. 01001583 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-001-583 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-001-583 Supplier MilliporeSigma Supelco Supplier No. M0301ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Certified reference material

Morphine is a very potent opiate analgesic drug used to relieve both acute and chronic severe pain. Sold under trade names including MS Contin™, Kadian™, Oramorph™, and Roxanol, morphine is also a commonly abused recreational drug due to its widespread availability and high potential for addiction. This certified analytical reference standard is suitable for a variety of GC/MS or LC/MS applications from pain prescription monitoring, clinical toxicology and urine drug testing to pharmaceutical research and forensic analysis.
TRUSTED_SUSTAINABILITY

Specifications

CAS 67-56-1, 57-27-2
Linear Formula C17H19NO3
UN Number UN 1230 - class 3 - PG 2 - Methanol, solution
Concentration 100 μg/mL in methanol
Formula Weight 285.34
Packaging Ampule
Quantity 1 mL
Flash Point 9.7°C (49.5°F) - closed cup

CERILLIANT is a registered trademark of Cerilliant Corporation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.